Learn More
Description
- EDTA added to trypsin solutions to enhance activity
- Contains 0.5g/L Trypsin 1:250; 0.2g/L Versene (EDTA)
- Manufactured with irradiated trypsin
- Porcine parvovirus tested and mycoplasma screened
Specifications
Specifications
| Content And Storage | -10° to -20°C |
| Product Type | Cell Culture Dissociation Reagent |
| Sterility | Sterile |
| pH | 7.1 to 7.9 |
| Quantity | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.