Learn More
Lonza™ BioWhittaker™ DPBS (1X) w/Calcium and Magnesium, Glutamine and Sodium Pyruvate
Shop All Lonza, Inc ProductsSpecifications
Specifications
| Content And Storage | 15° to 30°C |
| Product Type | Dulbecco's Phosphate Buffered Saline |
| pH | 7 to 7.6 |
| Quantity | 1 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.