Learn More
Description
- Used to provide buffer system to maintain cell culture media in physiological pH range of 7.0 to 7.6
- Serves as irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for limited period of time
Specifications
Specifications
| Content And Storage | 15° to 30°C |
| Product Type | Dulbecco's Phosphate Buffered Saline |
| Without Additives | No Calcium, No Magnesium |
| pH | 7 to 7.6 |
| Quantity | 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.