Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Ham's F-10 Medium

Catalog No. BW12618F Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW12-618F 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BW12-618F Supplier Lonza™ Supplier No. 12618F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Originally designed for low density, serum-free growth of Chinese Hamster Ovary (CHO) cells
  • Also used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells

TRUSTED_SUSTAINABILITY

Specifications

Product Type Ham's F10
Sterility Sterile
Sterilization Method Sterile-filtered through two consecutive 0.2 μm filters
Quantity 500 mL
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.