Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Ham's F-12 Medium

Catalog No. BW12615F Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW12-615F 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BW12-615F Supplier Lonza™ Supplier No. 12615F

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for low density, serum-free growth of Chinese Hamster Ovary (CHO) cells. BioWhittaker™ Ham's F-12 Medium has been used for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells and rat, chicken and rabbit embryos.

  • Nutrient mixture designed to cultivate a wide variety of mammalian and hybridoma cells when used with serum in combination with hormones and transferrin
    • F-12 medium is based on Ham’s F-10 medium with increased concentrations of choline, inositol, putrescine and many amino acids
    • Manufactured in accordance with cGMP regulations
    • Formulations used for standard liquid media products are those recommended by the Tissue Culture Association
TRUSTED_SUSTAINABILITY

Specifications

Product Type Ham's F-12
Sterility Sterile
Sterilization Method Sterile-filtered through two consecutive 0.2 μm filters
Quantity 500 mL
pH 7.07 to 7.4
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.