Learn More
Description
- Used in open vessels incubated under atmospheric conditions
- Behaves as zwitterion
- Sodium chloride concentration of HEPES buffered medium has been reduced to maintain same Osmolality of original media formulations
- Manufactured in accordance with cGMP regulations
- Cell culture-tested to assure compatibility
Specifications
Specifications
| Content And Storage | 15° to 30°C |
| Product Type | HEPES Buffer |
| Sterility | Sterile |
| pH | 6.98 to 7.3 |
| Quantity | 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.