Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ McCoy's 5A Medium (Modified)

Catalog No. BW12168F
Change view
Click to view available options
pH:
6.89 to 7.2
7.03 to 7.51
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
BW12-168F 7.03 to 7.51
BW12-688F 6.89 to 7.2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BW12-168F Supplier Lonza™ Supplier No. 12168F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

General purpose medium for primary and established cell lines. BioWhittaker™ McCoy's 5A Medium (Modified) is manufactured in accordance with cGMP regulations.

  • Formulated for the nutritional requirements of Walker 256 carcinoma
  • Unique components include asparagine, 3X normal glucose, amino acids (all L-isomers) and increased folic acid
  • Manufactured in accordance with cGMP regulations
  • Formulations used for standard liquid media products are those recommended by Tissue Culture Association
TRUSTED_SUSTAINABILITY

Specifications

Product Type McCoy's 5A
Sterility Sterile
Sterilization Method Sterile-filtered through two consecutive 0.2 μm filters
Quantity 500 mL
pH 7.03 to 7.51
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.