Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Medium 199 with Earle's BSS

Catalog No. BW12119F Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
500 mL
With Additives:
Glutamine, HEPES
Glutamine, Sodium Bicarbonate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives
BW12-119F 500 mL Glutamine, Sodium Bicarbonate
BW12-117F 500 mL Glutamine, HEPES
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BW12-119F Supplier Lonza™ Supplier No. 12119F

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Uses include virology and vaccine production
  • Components include purines (adenine, guanine, xanthine, hypoxanthine), pyrimdines (thymine, uracil) and fat soluble compounds

TRUSTED_SUSTAINABILITY

Specifications

Product Type M199/EBSS
Sterility Sterile
Sterilization Method Sterile-filtered through two consecutive 0.2 μm filters
With Additives Glutamine, Sodium Bicarbonate
Quantity 500 mL
pH 6.9 to 7.3
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.