Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ MEM Eagle with Hanks' BSS

Catalog No. BW12127F Shop All Lonza, Inc Products
Change view
Click to view available options
pH:
6.92 to 7.4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
BW12-127F 6.92 to 7.4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BW12-127F Supplier Lonza™ Supplier No. 12127F

missing translation for 'validMainframeMsg'

MEM Eagle is suitable for a diverse spectrum of mammalian cell types.

TRUSTED_SUSTAINABILITY

Specifications

Product Type MEM Eagle/HBSS
Sterility Sterile
Sterilization Method Sterile-filtered through two consecutive 0.2 μm filters
Without Additives No Glutamine
Quantity 500 mL
pH 6.92 to 7.4
Content And Storage 15° to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.