Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ MEM Eagle Vitamin Mixture (100X)

Catalog No. BW13607C Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW13-607C 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BW13-607C Supplier Lonza™ Supplier No. 13607C

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured in accordance with cGMP regulations

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -10° to -20°C
Product Type Vitamin Supplement
Sterility Sterile
pH 4.53 to 7.05
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.