Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Sodium Pyruvate Solution (100mM)

Catalog No. BW13115E Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW13-115E 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BW13-115E Supplier Lonza™ Supplier No. 13115E

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Added to cell culture as an additional source of energy since it is a key intermediate in the production of the high energy intermediate ATP within the cell BioWhittaker™ Sodium Pyruvate Solution (100mM) is manufactured in accordance with cGMP regulations.

  • Sodium Pyruvate Solution may help cells that are deficient in the nine enzymatic steps that break glucose down to pyruvate.
  • May also assist in “jump starting” cells in the citric acid cycle and thus help low density cells grow and produce energy in the form of ATP
  • Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Form Liquid
Product Type Sodium Pyruvate
Sterility Sterile
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.