Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Versene™ (EDTA), 0.02%

Catalog No. BW17711E Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW17711E 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BW17711E Supplier Lonza™ Supplier No. 17711E

missing translation for 'validMainframeMsg'

It is a chelating agent used to lift some cell types from the culture vessel surface into suspension

  • Manufactured in accordance with cGMP regulations
  • Cell culture tested to assure compatibility

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type Cell Culture Dissociation Reagent
Without Additives No Calcium, No Magnesium
pH 7.08 to 7.5
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.