Learn More
Description
• Ladders are prepared from five highly purified DNA fragments in perfect relative mass equivalence
• Reverse QuantLadders reverse the mass equivalence order of the QuantLadder
• SimplyLoad™ ready-to-load ladders contain loading buffer; other ladders are supplied with separate loading buffer
• Store at 2° to 8°C for up to 12 months
Specifications
Specifications
| Buffer | TE |
| Concentration | 100 μg/mL |
| Form | Liquid |
| Formulation | Includes a vial of 6X sample loading buffer 1mL total volume 15% w/v Ficoll™ 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene Cyanol FF 30mM EDTA |
| For Use With (Application) | Quantitation of DNA within ±1% accuracy |
| No. of Bands | 5 |
| Quantity | 125 μL |
| Storage Requirements | 2° to 8°C |
| Product Type | DNA QuantLadder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.