Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ DNA QuantLadders

Catalog No. p-2796845
Click to view available options
Product Type:
DNA QuantLadder

Provide convenient, visual quantitation of DNA within ±1% accuracy

•    Ladders are prepared from five highly purified DNA fragments in perfect relative mass equivalence
•    Reverse QuantLadders reverse the mass equivalence order of the QuantLadder
•    SimplyLoad™ ready-to-load ladders contain loading buffer; other ladders are supplied with separate loading buffer
•    Store at 2° to 8°C for up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

Buffer TE
Concentration 100 μg/mL
Product Type DNA QuantLadder
Form Liquid
Formulation Includes a vial of 6X sample loading buffer 1mL total volume 15% w/v Ficoll™ 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene Cyanol FF 30mM EDTA
For Use With (Application) Quantitation of DNA within ±1% accuracy
No. of Bands 5
Quantity 125 μL
Storage Requirements 2° to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.