Description
Includes
FlashGel RNA Cassettes, 1.2% agarose (9 Pk.), 12 + 1 well single tier; Formaldehyde Sample Buffer; FlashGel RNA Marker; and AccuGENE™ Molecular Biology Water
- Provide nearly instant results
- Load samples, watch bands migrate and get data in as little as two minutes
- Needs no gel preparation, band excision, purification, and UV light
- Complete separation, recovery and documentation safely at the bench
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
| For Use With (Application) | Analysis of total RNA, quick checks of native RNA and checking RNA degradation and mRNA purity |
| Kit Contents | FlashGel™ RNA Cassettes 1.2% Agarose 12 + 1 well, Formaldehyde Sample Buffer, FlashGel™ RNA Marker and AccuGENE Molecular Biology Water |
| Quantity | 9 Pack |