Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ FlashGel™ System DNA Markers

Catalog No. bma50473
Change view
Click to view available options
No. of Bands:
6
8
For Use With (Application):
1.2% FlashGel DNA Cassette
2.2% FlashGel DNA Cassette
Double-tier FlashGel DNA Cassette
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Bands For Use With (Application)
BMA50473 8 1.2% FlashGel DNA Cassette
BMA57033 8 2.2% FlashGel DNA Cassette
BMA57034 6 Double-tier FlashGel DNA Cassette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Do not exceed 5μL per well load volume on FlashGel cassettes
Catalog No. BMA50473 Supplier Lonza™ Supplier No. 50473
Only null left
Add to Cart
Add to Cart
Do not exceed 5μL per well load volume on FlashGel cassettes

Provide accurate size estimation of DNA fragments. FlashGel™ System DNA Markerswere designed to optimize performance of the FlashGel™ system and come ready-to-use.

  • Ready-to-load molecular weight standards
  • Separates DNA in real time: 2 to 7 minutes
  • Clean, sharp separation and straight, uniform sample lanes
  • Watch DNA migration as it happens: Monitor gel runs right at the bench, without UV light
  • Highly sensitive system allows a reduction in DNA levels, saving time and money
  • Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
  • Detect <0.1ng DNA or <10ng total RNA
  • Recommended for FlashGel DNA cassettes

 

FlashGel 1.2% DNA Marker

  • Sizes: 100/200/300/500/800/1250/2000/4000bp
  • Recommended for 1.2% FlashGel DNA cassettes
FlashGel 2.2% DNA Marker
  • Sizes: 50/100/150/200/300/500/800/1500bp
  • Recommended for 2.2% FlashGel DNA cassettes
FlashGel Double Tier DNA Marker
  • Sizes: 100/300/500/800/1500/3000bp
  • Recommended for double tier FlashGel DNA cassettes

 

 

  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Formulation 5μL Load 1:5 dilution
For Use With (Application) 1.2% FlashGel DNA Cassette
No. of Bands 8
Quantity 500 μL
Storage Requirements 4°C
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.