Learn More
Description
- Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
- Recover samples directly, without band excision or purification
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
| Concentration | 2.2% |
| Separation Range | 10 to 1000 bp |
| Wells | 8 + 1-well |
| Formulation | 2.2% Agarose |
| For Use With (Application) | Subsequent re-amplification, cloning or other techniques |
| Kit Contents | Agarose Gel, Stain and Buffer |
| Material | Agarose |
| Quantity | 9 Pack |
| Storage Requirements | 18° to 26°C |
| Length (English) Cassette | 4.5 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.