Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ FlashGel™ System Recovery Cassettes

Catalog No. p-4684349
Change view
Click to view available options
Separation Range:
10 to 1000 bp
50 to 10,000 bp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Separation Range
BMA57051 50 to 10,000 bp
BMA57022 10 to 1000 bp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMA57051 Supplier Lonza™ Supplier No. 57051
Only null left
Add to Cart
Add to Cart

Eliminates agarose gel preparation, band excision and purification. Lonza™ FlashGel™ - System Recovery Cassettes deliver recovery, free from inhibitors and UV-induced damage, in a simple 5-10 minute protocol.

  • Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
  • Recover samples directly, without band excision or purification
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.2%
Separation Range 50 to 10,000 bp
Wells 8 + 1-well
Formulation 1.2% Agarose
For Use With (Application) Subsequent re-amplification, cloning or other techniques
Kit Contents Agarose Gel, Stain and Buffer
Material Agarose
Quantity 9 Pack
Storage Requirements 18° to 26°C
Length (English) Cassette 4.5 in.
Length (English) Gel 2.7 in.
Length (Metric) Cassette 11.5 cm
Length (Metric) Gel 7 cm
Width (English) Cassette 4.2 in.
Width (Metric) Cassette 10.7 cm
Width (Metric) Gel 8.4 cm
Height (English) Cassette 0.6 in.
Height (Metric) Cassette 1.7 cm
Product Type System Recovery Cassettes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.