Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ FlashGel™ System RNA Cassettes

Catalog No. bma57027
Change view
Click to view available options
Wells:
12 + 1-well
16 + 1-well
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
BMA57027 12 + 1-well
BMA57028 16 + 1-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMA57027 Supplier Lonza™ Supplier No. 57027
Only null left
Add to Cart
Add to Cart

Cassettes optimized for RNA. FlashGel™ System RNA Cassettes are recommended for verification and analysis of total RNA, quick checks of native RNA, and checking RNA degradation and mRNA purity.

Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains and hazardous reagents
  • Casettes fully enclose the gel, stain and running buffer
  • Protect samples from contaminating RNases
  • Designed for performance and purity, and are guaranteed RNase-free
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.2%
Separation Range 0.5 to 9 kb
Wells 12 + 1-well
Formulation 1.2% Agarose
For Use With (Application) Analysis of total RNA, quick checks of native RNA and checking RNA degradation and mRNA purity
Kit Contents Agarose Gel, Stain and Buffer
Material Agarose
Quantity 9 Pack
Length (English) Cassette 4.5 in.
Length (English) Gel 2.7 in.
Length (Metric) Cassette 11.5 cm
Length (Metric) Gel 7 cm
Width (English) Cassette 4.2 in.
Width (Metric) Cassette 10.7 cm
Width (Metric) Gel 8.4 cm
Height (English) Cassette 0.6 in.
Height (Metric) Cassette 1.7 cm
Product Type RNA Cassettes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.