Learn More
Description
Designed to optimize performance of the FlashGel System (Cat. No. BMA57067): optimized for the unique requirements of RNA. Provides consistent results in gene expression, Northern analysis, cDNA library construction and cDNA labeling for microarrays.
- 50μL
- Band sizes: 0.5/1/1.5/3/5/9kbp
- Results in 30 minutes or less
- Detect <10ng RNA per band
- Avoid hazardous reagents and contaminating RNases
- Cassettes contain precast, pre-stained agarose gels and buffer: no need for gel preparation, buffer addition or gel staining
- Clean, enclosed system, provided ready-to-load
- Denature FlashGel RNA markers with Formaldehyde sample Buffer (Cat. No. BW50571) for 5 minutes at 65°C (149°F)
- For native RNA, combine with FlashGel loading dye (Cat. No. BMA50462)
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
- Verification and analysis of total RNA
- Quick checks of native RNA
- Checking for RNA degradation and mRNA purity
Specifications
Specifications
| Concentration | 1 μg/mL |
| Form | Liquid |
| For Use With (Application) | Accurate size estimation of DNA fragments |
| No. of Bands | 6 |
| Quantity | 50 μL |
| Storage Requirements | −80°C |
| Product Type | RNA Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.