Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ GelStar™ Nucleic Acid Gel Stain

Catalog No. bma50535
Change view
Click to view available options
Quantity:
2 x 250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMA50535 2 x 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA50535 Supplier Lonza™ Supplier No. 50535
Only null left
Add to Cart
Add to Cart

Use to detect small quantities of DNA and RNA. GelStar™ Nucleic Acid Gel Stain exhibits exceptional signal-to-noise ratio with minimal background.

  • Detects as little as 20pg of DNA or 3ng of RNA
  • Exhibits exceptional signal-to-noise ratio with minimal background
  • Compatible with agarose or polyacrylamide gels
  • Can be added to gel before casting or used for post-electrophoresis staining
  • Can be visualized with a standard 300nm UV transilluminator or the Dark Reader™ transilluminator
  • Ideal alternative to silver staining
  • For research use only

Product licensed by Cambrex from Molecular Probes, Inc. For research and quality control use only.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Concentration 10,000X
Form Liquid
For Use With (Application) Agarose or Polyacrylamide gels
Quantity 2 x 250 μL
Storage Requirements −20°C
Product Type DNA/RNA Stain
Stain Type Nucleic Acid Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.