Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Glyoxal Sample Buffer

Catalog No. BMA50560 Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
1.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMA50560 1.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA50560 Supplier Lonza™ Supplier No. 50560

missing translation for 'validMainframeMsg'

Specifications

Formulation Glyoxal and DMSO in MOPS Buffer
For Use With (Application) RNA Electrophoresis
Packaging Type Vial
Quantity 1.7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.