Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ IsoGel™ IEF Plate Accessories

Catalog No. BMA56014 Shop All Lonza, Inc Products
Change view
Click to view available options
Product Type:
Accessory Pack
Blotting paper
Electrophoresis Accessory Bulk Pack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BMA56014 Accessory Pack
BMA56007 Blotting paper
BMA56010 Electrophoresis Accessory Bulk Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BMA56014 Supplier Lonza™ Supplier No. 56014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessories for precast agarose IEF plates. Blotting paper and accessory packs.

  • Blotting paper is for use with precast agarose IEF plates
  • Accessory pack includes masks, 100mm and 125mm wicks and blotting paper
  • Accessory bulk pack includes 125mm wicks and blotting paper
TRUSTED_SUSTAINABILITY

Specifications

Separation Type Isoelectric Focusing
Form Dried
For Use With (Application) Isoelectric focusing, Antibody separation and analysis
Storage Requirements 18° to 26°C
Product Type Accessory Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.