Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ IsoGel™ IEF Plates

Catalog No. BMA56015 Shop All Lonza, Inc Products
Change view
Click to view available options
pH Range:
3 to 10
6 to 10.5
7 to 11
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
BMA56015 3 to 10
BMA56018 6 to 10.5
BMA56024 7 to 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BMA56015 Supplier Lonza™ Supplier No. 56015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Precast agarose IEF plates provide rapid and reproducible isoelectric focusing of proteins up to 2000kDa

  • Specifically designed for antibody analysis of
  • Each gel is supported on GelBond™ film for easy handling and dimensional stability
  • Gel size is compatible with most IEF chambers
  • 125 x 100mm

Isoelectric focusing, antibody separation and analysis

Order Info

Accessories packs (Cat. No. BMA56014 and BMA56010) and blotting paper (BMA56007) available separately.

TRUSTED_SUSTAINABILITY

Specifications

pH Range 3 to 10
Separation Type Isoelectric Focusing
Form Solid
For Use With (Application) Isoelectric focusing, Antibody separation and analysis
Storage Requirements 2° to 8°C
Product Type Gel Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.