Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Latitude™ Precast Agarose Midigels

Catalog No. BMA57231 Shop All Lonza, Inc Products
Change view
Click to view available options
Separation Range:
100 to ≥2000 bp
300 to ≥8000 bp
Wells:
2 x 20-well
20-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Separation Range Wells
BMA57231 100 to ≥2000 bp 2 x 20-well
BMA57230 300 to ≥8000 bp 2 x 20-well
BMA57220 300 to ≥8000 bp 20-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BMA57231 Supplier Lonza™ Supplier No. 57231
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Efficiently resolve DNA ranging from 10bp to more than 10kb. Latitude™ Precast Agarose Midigels are designed for accurate DNA fragment separation for most any gel electrophoresis application.

  • Ideal for checking restriction digests and PCR products
  • Quality and performance tested to ensure reproducible results every time
  • Prestained with ethidium bromide
  • Compatible with common midichambers, multichannel pipets and alternate-well microtiter plate spacing

Note:ethidium bromide is a powerful mutagen; gloves should be worn when opening pouches and handling gels and solutions containing ethidium bromide

TRUSTED_SUSTAINABILITY

Specifications

Buffer TBE
Color Clear
Concentration 2%
Format Precast Gel
Separation Range 100 to ≥2000 bp
Wells 2 x 20-well
Dnase Free Yes
Formulation 2% SeaKem™ LE Plus Agarose, 1X TBE, Prestained with 0.5μg/mL of Ethidium Bromide
For Use With (Application) Designed for accurate DNA fragment separation for most any Gel electrophoresis application
Material Agarose
Quantity 8 Gels
RNase Free Yes
Storage Requirements 18° to 26°C
Thickness (English) 0.23 in.
Thickness (Metric) 0.6 cm
Volume Per Well 10 to 12 μL
Product Type Agarose Gel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.