Learn More
Description
Includes
- Include
- Long Ranger Gel Solution
- 1X TBE
- Urea
- APS
- TEMED
- Filter
All reagents necessary for casting a single gel are premeasured and packaged in a ready-to-use pouch. The reagents are mixed in the pouch and cast directly from the pouch through an internal filter.
- Compatible with the ABI Prism 373, ABI Prism 377, ALFexpress, or LI-COR automated DNA sequencers
- Eliminate the need to measure individual reagents
- Ideal for DNA and RNA sequencing, STR analysis, identity testing, genotyping, and differential display
- Store at 18° to 24°C
Specifications
Specifications
| Product Type | Agarose Gel |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.