Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ PAGEr™ Gold 4% Tris-Glycine, Mini Protein Gel, 17-well

Catalog No. BMA59560 Shop All Lonza, Inc Products
Change view
Click to view available options
Wells:
17-well
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
BMA59560 17-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA59560 Supplier Lonza™ Supplier No. 59560

missing translation for 'validMainframeMsg'

Sharp, reliable, easy-to-use Tris-Glycine protein gels

PAGEr Gold Precast Gels are easy-to-use protein minigels that offer sharp resolution and consistent protein transfer. Choose from multiple options for denatured or native protein separation over a wide molecular weight range, in an array of well configurations in both 9 × 10cm (see BMA58517 through BMA58524) and 10 × 10cm sizes to fit most popular chambers.

  • Polyacrylamide minigels cast with 4% stacking gel for vertical protein electrophoresis
  • Provide easy, accurate separation of biomolecules over wide molecular-weight range
  • Produce crisp, razor-sharp resolution 5kDa - 250kDa
  • Tris-Glycine buffer - Traditional Laemmli separation
  • Marked sample lanes for easy loading and a simple twist-open design
  • 8+1- and 17-well configurations are compatible with multichannel pipetters
  • 2D-well configuration is designed for antibody testing and two-dimensional electrophoresis using a 7cm IPG strip

  • Fit most vertical minigel boxes
  • Packaged in a unique easy-to-open gold plastic cassette
  • Available with a variety of polyacrylamide concentrations and well configurations

Certifications

ISO 9001:2000

TRUSTED_SUSTAINABILITY

Specifications

Cassette Material Plastic
Certifications/Compliance ISO 9001:2000
Gel Percentage 4%
Gel Size Mini
Gel Thickness 0.55 cm
Gel Type Tris-Glycine
Separation of Protein
Separation Range 5 to 250 kDa
Separation Type Denaturing, Native
Wells 17-well
For Use With (Application) 2D Electrophoresis, Denaturing and Native Protein Electrophoresis, Western Blotting
Quantity 10/Pk.
Well Design 2D Well
Length (Metric) 10 cm
Width (Metric) 10 cm
Product Type Gradient Page
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.