Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ PBS EDTA

Catalog No. BWBE02017F Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BWBE02017F 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BWBE02017F Supplier Lonza™ Supplier No. BE02017F

missing translation for 'validMainframeMsg'

Used as a buffer in procedures generating dendritic cells from monocytes. BioWhittaker™ PBS EDTA is manufactured in accordance with cGMP regulations.

 

  • For further manufacturing or laboratory use
  • Molarity of EDTA is 1.236mM
  • Quantity: 500mL

 

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.