Learn More
Description
- Stain less than 10 minutes
- 1-step process
- Non-toxic
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | Mass spectrometry, silver staining, PVDF membranes |
| Quantity | 25 mL |
| Product Type | Protein Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.