Learn More
Description
- Ready to use
- RNase free
- Free of denaturants, eliminating problems associated with staining gels containing formaldehyde
- Suitable for a variety of applications, including Northern blotting and analysis of RNA transcripts
- Compatible with most common mini chambers
- Available with a separate container of glyoxal buffer
Specifications
Specifications
| Buffer | MOPS |
| Concentration | 1.25% |
| Format | Precast Gel |
| Wells | 20-well |
| Dnase Free | Yes |
| Formulation | 1.25% SeaKem™ Gold Agarose, 1X MOPS |
| For Use With (Application) | Northern blotting and analysis of RNA transcripts |
| Material | Agarose |
| Quantity | 20 Gels |
| RNase Free | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.