Description
- DNA recovered from SeaKem™ GTG™ Agarose gels may be reliably digested and ligated
- Extensively performance tested to ensure complete compatibility with routine molecular biology techniques
Specifications
Specifications
| Chemical Name or Material | SeaKem GTG Agarose |
| Quantity | 25 g |