Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeaKem™ HE Agarose, Lonza™

Catalog No. BMA50020
Change view
Click to view available options
Quantity:
25 g
125 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMA50020 125 g
BMA50021 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BMA50020 Supplier Lonza Inc Supplier No. 50020
Add to Cart
Add to Cart
This item is not returnable. View return policy

  • For immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CEIP), and serum protein electrophoresis
  • High EEO (0.23 –0.26)
  • Standard gelling temperature agarose

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material SeaKem HE Agarose
Quantity 125 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.