Description
- For immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CEIP), and serum protein electrophoresis
- High EEO (0.23 –0.26)
- Standard gelling temperature agarose
Specifications
Specifications
| Chemical Name or Material | SeaKem HE Agarose |
| Quantity | 25 g |