Description
- Used in counterimmunoelectrophoresis and crossed immunoelectrophoresis
- High gelling temperature (42°C ± 1.5°C)
- High gel strength (≥1000g/cm2) and clarity
Specifications
Specifications
| Chemical Name or Material | SeaKem HGT Agarose |
| Quantity | 25 g |