Learn More
Description
• Consist of RNA transcripts in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9kb
• Can be denatured with standard procedures
• Can be visualized on Northern blots with labeled lambda sequence
• Can be detected with ethidium bromide or nucleic acid stains
• Store at -20°C for up to six months or at -80°C for up to 24 months
Specifications
Specifications
| Color | Blue, Cyan |
| Concentration | 60 ng/μL |
| Form | Liquid |
| Formulation | 50% GC content |
| For Use With (Application) | Gel electrophoresis |
| Increments | 100bp and 1000bp |
| No. of Bands | 21 |
| Quantity | 500 μL |
| Storage Requirements | 2° to 8°C |
| Product Type | DNA Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.