Learn More
Description
Does not interfere with subsequent immunodetection.
Product licensed by Cambrex from Molecular Probes, Inc. For research and quality control use only.
Specifications
Specifications
| Color | Ruby |
| Form | Liquid |
| For Use With (Application) | Fluorogenic, chemiluminescent and colorimetric detection techniques |
| Quantity | 200 mL |
| Product Type | Protein Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.