Learn More
Lonza™ Williams' Medium E, without L-glutamine
Catalog No. BWBE02019F Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
BWBE02019F 500 mL
1 options
Catalog No. BWBE02019F Supplier Lonza™ Supplier No. BE02019F
For in vitro diagnostic use
  • Formulation used for this standard liquid media product is recommended by the Tissue Culture Association
  • Manufactured in accordance with cGMP regulations
For further manufacturing or laboratory use
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 mL