Learn More
Sigma Aldrich Fine Chemicals Biosciences LookOut Mycoplasma qPCR Detection Kit, 1 kit
Catalog No. NC1668597
Encompass
The LookOut Mycoplasma PCR Detection Kit utilizes the polymerase chain reaction (PCR), which is established as the method of choice for highest sensitivity in the detection of Mycoplasma and Acholeplasma contamination in cell cultures and other cell culture derived biologicals.
Catalog No. NC1668597 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MP0040A1KT
The LookOut Mycoplasma PCR Detection Kit utilizes the polymerase chain reaction (PCR), which is established as the method of choice for highest sensitivity in the detection of Mycoplasma and Acholeplasma contamination in cell cultures and other cell culture derived biologicals.