Learn More
Instech Loop Connector for 2 Channel Rat Vascular Access Button™, Primer
SDP
Catalog No. NC2816942
Encompass
The Instech VABR2L is a two-channel loop connector designed for rats using the Vascular Access Button™ system. This specialized shunt links the red and white ports of a 2-channel button to create a continuous extracorporeal circuit, typically between a bile duct catheter and a duodenal catheter. By allowing bile to circulate naturally during recovery or between sampling intervals, the loop maintains the animal's physiological homeostasis and prevents bile salt depletion. The low-profile connector attaches magnetically, providing a secure yet low-stress connection that allows for pair or group housing when used with a protective cap.
Features
  • Product Type: 2-channel loop connector (shunt)
  • Compatibility: 2-channel Rat VAB™ (VABR2B)
  • Application: Bile duct to duodenum bypass
  • Connection: Magnetic quick-connect
  • Benefit: Maintains enterohepatic circulation
  • Housing: Compatible with VABRC protective caps
Catalog No. NC2816942 Supplier Instech Supplier No. VABR2L
The Instech VABR2L is a two-channel loop connector designed for rats using the Vascular Access Button™ system. This specialized shunt links the red and white ports of a 2-channel button to create a continuous extracorporeal circuit, typically between a bile duct catheter and a duodenal catheter. By allowing bile to circulate naturally during recovery or between sampling intervals, the loop maintains the animal's physiological homeostasis and prevents bile salt depletion. The low-profile connector attaches magnetically, providing a secure yet low-stress connection that allows for pair or group housing when used with a protective cap.
Features
  • Product Type: 2-channel loop connector (shunt)
  • Compatibility: 2-channel Rat VAB™ (VABR2B)
  • Application: Bile duct to duodenum bypass
  • Connection: Magnetic quick-connect
  • Benefit: Maintains enterohepatic circulation
  • Housing: Compatible with VABRC protective caps