Learn More
Instech Loop Connector for 3 Channel Rat Vascular Access Button™ (white to blue), Primer
SDP
Catalog No. NC2816943
Encompass
The Instech VABR3L is a sterile loop connector designed for the three-channel rat Vascular Access Button™, enabling bile duct–to–duodenum catheterization by linking the white and blue PinPorts. It preserves access to the red port for sampling or infusion and integrates seamlessly with the VABR3B/22 button system.
Features
  • Function: Connects white → blue channels to create a closed bile circulation loop
  • Application: Supports bile duct / duodenum catheterization workflows
  • Compatibility: Designed for the Instech VABR3B/22 3‑channel Vascular Access Button™
  • Access: Includes PinPort™ primer for syringe connection using PNP3M injector
  • Sterility: Supplied sterile (EtO)
  • Port Availability: Red port remains available for infusion or sampling
  • Quantity: 1 loop connector
Catalog No. NC2816943 Supplier Instech Supplier No. VABR3L
The Instech VABR3L is a sterile loop connector designed for the three-channel rat Vascular Access Button™, enabling bile duct–to–duodenum catheterization by linking the white and blue PinPorts. It preserves access to the red port for sampling or infusion and integrates seamlessly with the VABR3B/22 button system.
Features
  • Function: Connects white → blue channels to create a closed bile circulation loop
  • Application: Supports bile duct / duodenum catheterization workflows
  • Compatibility: Designed for the Instech VABR3B/22 3‑channel Vascular Access Button™
  • Access: Includes PinPort™ primer for syringe connection using PNP3M injector
  • Sterility: Supplied sterile (EtO)
  • Port Availability: Red port remains available for infusion or sampling
  • Quantity: 1 loop connector