Learn More
Instech Loop Connector for 4 Channel rat Vascular Access Button™ (white to blue), Primer
SDP
Catalog No. NC2816950
Encompass
The Instech VABR4L is a loop connector designed specifically for the 4-channel Rat Vascular Access Button™. This specialized connector links two of the button's ports to create a continuous fluid circuit, which is essential for studies involving bile duct to duodenum bypass. By allowing the bile to circulate naturally between samples, the loop maintains the animal's physiological balance and prevents the depletion of bile salts. When a sample or infusion is required, the loop is temporarily replaced with a mating tether, ensuring a closed and aseptic system throughout the duration of the longitudinal study.
Features
  • Product Type: Loop connector
  • Compatibility: 4-channel Rat VAB™
  • Application: Bile duct / Duodenal bypass
  • Benefit: Maintains physiological homeostasis
  • Design: Low-profile, magnetic snap-fit
  • Material: Medical-grade, biocompatible fluid path
Catalog No. NC2816950 Supplier Instech Supplier No. VABR4L
The Instech VABR4L is a loop connector designed specifically for the 4-channel Rat Vascular Access Button™. This specialized connector links two of the button's ports to create a continuous fluid circuit, which is essential for studies involving bile duct to duodenum bypass. By allowing the bile to circulate naturally between samples, the loop maintains the animal's physiological balance and prevents the depletion of bile salts. When a sample or infusion is required, the loop is temporarily replaced with a mating tether, ensuring a closed and aseptic system throughout the duration of the longitudinal study.
Features
  • Product Type: Loop connector
  • Compatibility: 4-channel Rat VAB™
  • Application: Bile duct / Duodenal bypass
  • Benefit: Maintains physiological homeostasis
  • Design: Low-profile, magnetic snap-fit
  • Material: Medical-grade, biocompatible fluid path