Learn More
Instech Loop connector for 2 channel mouse VAB™/VAH™, primer
SDP
Catalog No. NC3432939
Encompass
The Instech VABM2L is a loop connector designed for the two‑channel mouse Vascular Access Button™, enabling the red and white channels to be linked for applications such as bile circulation. By providing a simple, sterile connection between the two fluid paths, the VABM2L supports closed‑system operation, maintains catheter patency, and allows specialized workflows requiring intra-animal recirculation.
Features
  • Function: Loop connector for 2‑channel mouse VAB
  • Application: Enables channel-to-channel fluid recirculation (e.g., bile circulation)
  • Compatibility: VABM2 series mouse Vascular Access Buttons
  • Access: Connects red and white PinPort™ channels
  • Design: Sterile, closed‑system accessory
  • Use Case: Supports infusion/sampling models requiring internal looping
Catalog No. NC3432939 Supplier Instech Supplier No. VABM2L
The Instech VABM2L is a loop connector designed for the two‑channel mouse Vascular Access Button™, enabling the red and white channels to be linked for applications such as bile circulation. By providing a simple, sterile connection between the two fluid paths, the VABM2L supports closed‑system operation, maintains catheter patency, and allows specialized workflows requiring intra-animal recirculation.
Features
  • Function: Loop connector for 2‑channel mouse VAB
  • Application: Enables channel-to-channel fluid recirculation (e.g., bile circulation)
  • Compatibility: VABM2 series mouse Vascular Access Buttons
  • Access: Connects red and white PinPort™ channels
  • Design: Sterile, closed‑system accessory
  • Use Case: Supports infusion/sampling models requiring internal looping