Learn More
Med Vet International Fecal Loop Standard Size, 9in x 3/8in, J0147, Each
Catalog No. 502522334
Encompass_Preferred
Inexpensive plastic loop with slotted ends for quick and easy fecal samples. Reusable. This can easily be inserted into the rectum and a small amount of fecal material trapped in the loop. Can be cleaned, disinfected, and used repeatedly. Two convenient sizes available..
Catalog No. 50-252-2334 Supplier Med Vet International Supplier No. JOR147
Inexpensive plastic loop with slotted ends for quick and easy fecal samples. Reusable. This can easily be inserted into the rectum and a small amount of fecal material trapped in the loop. Can be cleaned, disinfected, and used repeatedly. Two convenient sizes available..