Learn More
World Precision Instrument Loop Filament-505104
SDP
Supplier:  World Precision Instrument 505104
Encompass
A glass puller is a laboratory instrument used to create fine, tapered glass micropipettes or microelectrodes. The choice of filament for the glass puller is crucial, as it directly influences the heating element's performance and, consequently, the quality of the pulled glass. Different types and shapes of filaments can be used, and the selection depends on factors such as the desired pipette. A loop filament is used in glass micropipette pullers. This filament design features a loop configuration with the ends of the wire coming together to form a closed loop. The loop filament is designed to influence the glass softening and pulling process in a way that creates specific shapes in the pulled capillaries. The loop filament has a single turn, and the ends of the wire meet to form a closed loop. The loop design allows for controlled and localized heating, influencing the glass softening and pulling process in a specific manner.
Catalog No. 50-253-9239