Learn More
Cole-Parmer Argos Technologies Brass Inoculating Loop and Needle Holder, 8"; Each
Catalog No. 501960656 Shop All Cole-Parmer Products
Encompass_Preferred
Reusable Inoculating Loops with handles are used to select, inoculate or transfer microbiological samples. Handles are insulated to protect the hand during use. Loops and Handles cool rapidly after sterilization. Loops and without handles are sold separately as models 04396-35 through -39. Needle without handle is available as model 04396-49.
Catalog No. 50-196-0656 Supplier Cole-Parmer Supplier No. 0439641
Reusable Inoculating Loops with handles are used to select, inoculate or transfer microbiological samples. Handles are insulated to protect the hand during use. Loops and Handles cool rapidly after sterilization. Loops and without handles are sold separately as models 04396-35 through -39. Needle without handle is available as model 04396-49.