Learn More
Med Vet International McKesson Inoculating Loop, Dark Color, 1 ?L, 20-Centimeter Length, 16-1RL, 1000/Case
Catalog No. 503044101
Encompass_Preferred
Inoculating Loop McKesson 1 µL Polystyrene Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Rigid feel media type Integrated handle Sterilized by gamma irradiation There are 25 calibrated inoculating loops in a tamper-evident resealable bag
Catalog No. 50-304-4101 Supplier Med Vet International Supplier No. 1175307CS
Inoculating Loop McKesson 1 µL Polystyrene Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Rigid feel media type Integrated handle Sterilized by gamma irradiation There are 25 calibrated inoculating loops in a tamper-evident resealable bag