Learn More
Med Vet International McKesson Inoculating Loop, Light Blue,10 µL, 20 cm Length, 16-10FL, 25/Bag
Catalog No. 503043225
Encompass_Preferred
Inoculating Loop McKesson 10 ?L High Impact Polystyrene Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Flexible feel media type Integrated handle Sterilized by gamma irradiation Light blue There are 25 calibrated inoculating loops in a tamper-evident resealable bag
Catalog No. 50-304-3225 Supplier Med Vet International Supplier No. 1175309BG
Inoculating Loop McKesson 10 ?L High Impact Polystyrene Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Flexible feel media type Integrated handle Sterilized by gamma irradiation Light blue There are 25 calibrated inoculating loops in a tamper-evident resealable bag