Learn More
Med Vet International McKesson Inoculating Loop, Light Green,1 ?L, 20-Centimeter Length, 16-1FL, 25/Bag
Catalog No. 503043273
Encompass_Preferred
Inoculating Loop McKesson 1 µL High Impact Polystyrene Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Flexible feel media type Integrated handle Sterilized by gamma irradiation Light green There are 25 calibrated inoculating loops in a tamper-evident resealable bag
Catalog No. 50-304-3273 Supplier Med Vet International Supplier No. 1175306BG
Inoculating Loop McKesson 1 µL High Impact Polystyrene Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Flexible feel media type Integrated handle Sterilized by gamma irradiation Light green There are 25 calibrated inoculating loops in a tamper-evident resealable bag