Learn More
Med Vet International McKesson Inoculating Loop with Needle, 1 µL, 20-Centimeter Length, 16-1FLWN, 1000/Case
Catalog No. 503044132
Encompass_Preferred
Inoculating Loop with Needle McKesson 1 µL ABS Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Flexible feel media type Integrated handle Sterilized by gamma irradiation There are 25 calibrated inoculating loops in a tamper-evident resealable bag
Catalog No. 50-304-4132 Supplier Med Vet International Supplier No. 1175308CS
Inoculating Loop with Needle McKesson 1 µL ABS Integrated Handle Sterile Disposables for collection and inoculation by streaking or puncturing methods Smooth construction facilitates uniform streaking of culture media Loop combinations have an integrated loop/needle design, eliminating the need for a separate needle Flexible feel media type Integrated handle Sterilized by gamma irradiation There are 25 calibrated inoculating loops in a tamper-evident resealable bag