Learn More
Med Vet International McKesson Inoculating Loop with Needle, 10 µL, 16-FLWN, 1000/Case
Catalog No. 503043287
Encompass_Preferred
Inoculating Loop with Needle McKesson 10 ?L ABS Integrated Handle Sterile Sterilized by gamma irradiation Disposable for collection and inoculation by streaking or puncturing methods Flexible feel Smooth construction facilitates uniform streaking of culture media Loop combinations feature integrated loop/needle design, eliminating the need for a separate needle
Catalog No. 50-304-3287 Supplier Med Vet International Supplier No. 1175311CS
 Restricted Item
Add to Cart
Add to Cart
Inoculating Loop with Needle McKesson 10 ?L ABS Integrated Handle Sterile Sterilized by gamma irradiation Disposable for collection and inoculation by streaking or puncturing methods Flexible feel Smooth construction facilitates uniform streaking of culture media Loop combinations feature integrated loop/needle design, eliminating the need for a separate needle